Assay Details
Target Gene Details
Entrez Gene ID: | 20304 |
Gene Name: | chemokine (C-C motif) ligand 5 |
Gene Aliases: |
MuRantes, RANTES, SISd, Scya5, TCP228 |
Location: |
Chr.11:83525778-83530518 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl5 | NM_013653.3 | 3 | 298 | NP_038681.2 |
AF065944.1 | 3 | 274 | AAC17511.1 | |
AF065945.1 | 3 | 274 | AAC17512.1 | |
AF065946.1 | 3 | 274 | AAC17513.1 | |
AF065947.1 | 3 | 274 | AAC17514.1 | |
AF128187.1 | 3 | 274 | AAF22528.1 | |
AK003101.1 | 3 | 299 | BAB22566.1 | |
AK158074.1 | 2 | 3385 | ||
AY722103.1 | AAU43779.1 | |||
BC033508.1 | 3 | 281 | AAH33508.1 | |
BE333205.1 | 1 | 67 | ||
CT010315.1 | CAJ18523.1 | |||
M77747.1 | 3 | 280 | AAA40029.1 | |
S37648.1 | 3 | 291 | AAB22330.1 |
More Information
Set Membership: |
Intragenic Exonic |