Assay Details
Target Gene Details
Entrez Gene ID: | 20302 |
Gene Name: | chemokine (C-C motif) ligand 3 |
Gene Aliases: |
AI323804, G0S19-1, LD78alpha, MIP-1alpha, MIP1-(a), MIP1-alpha, Mip1a, Scya3 |
Location: |
Chr.11:83647843-83649378 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl3 | NM_011337.2 | 3 | 465 | NP_035467.1 |
AA895994.1 | ||||
AK137525.1 | 1 | 1152 | ||
AK150590.1 | 3 | 445 | BAE29684.1 | |
AK150634.1 | 3 | 445 | BAE29723.1 | |
AK150698.1 | 3 | 445 | BAE29777.1 | |
AK151581.1 | 3 | 445 | BAE30521.1 | |
AK152648.1 | 3 | 445 | BAE31386.1 | |
AK153155.1 | 3 | 310 | BAE31765.1 | |
AK155058.1 | 1 | 1191 | ||
BC111443.1 | 3 | 389 | ||
BX524012.1 | 1 | 179 | ||
HG515534.1 | 2 | 229 | ||
J04491.1 | 3 | 411 | AAA40304.1 | |
M23447.1 | 3 | 442 | AAA40146.1 | |
X12531.1 | 3 | 434 | CAA31047.1 |
More Information
Set Membership: |
Intragenic Exonic |