Assay Details
Target Gene Details
Entrez Gene ID: | 20682 |
Gene Name: | SRY (sex determining region Y)-box 9 |
Gene Aliases: |
2010306G03Rik, AV220920, mKIAA4243 |
Location: |
Chr.11:112782210-112787757 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox9 | NM_011448.4 | 3 | 3629 | NP_035578.3 |
AK008535.1 | 1 | 323 | ||
AK049986.1 | 3 | 3615 | BAC34018.1 | |
AK220197.1 | 3 | 3599 | BAD90382.1 |