Assay Details
Target Gene Details
Entrez Gene ID: | 14257 |
Gene Name: | FMS-like tyrosine kinase 4 |
Gene Aliases: |
AI323512, Chy, Flt-4, VEGFR-3, VEGFR3 |
Location: |
Chr.11:49609679-49652739 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Flt4 | NM_008029.3 | 13 | 2047 | NP_032055.1 |
BC138346.1 | 13 | 2019 | ||
BC138348.1 | 13 | 2019 | ||
L07296.1 | 13 | 2000 | AAA40077.1 |
More Information
Set Membership: |
Intragenic Exonic |