Assay Details
Target Gene Details
Entrez Gene ID: | 12702 |
Gene Name: | suppressor of cytokine signaling 3 |
Gene Aliases: |
Cis3, Cish3, EF-10, Ef10, SSI-3, Ssi3 |
Location: |
Chr.11:117966079-117970341 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs3 | NM_007707.3 | 1 | 344 | NP_031733.1 |
AK047165.1 | 1 | 164 | BAC32977.1 | |
AK139241.1 | 1 | 165 | BAE23929.1 | |
AK150719.1 | 1 | 164 | BAE29797.1 | |
AK151761.1 | 1 | 164 | BAE30668.1 | |
AK152441.1 | 1 | 164 | BAE31222.1 | |
AK152468.1 | 1 | 164 | BAE31244.1 | |
AK152514.1 | 1 | 164 | BAE31277.1 | |
AK152686.1 | 1 | 164 | BAE31417.1 | |
AK155097.1 | 1 | 128 | ||
AK157708.1 | 1 | 150 | BAE34161.1 | |
AK159395.1 | 1 | 162 | BAE35049.1 | |
AK170406.1 | 1 | 162 | BAE41773.1 | |
AK172399.1 | 1 | 164 | BAE42985.1 | |
BC052031.1 | 1 | 140 | AAH52031.1 |
More Information
Set Membership: |
Intragenic Exonic |