Assay Details
Target Gene Details
Entrez Gene ID: | 28081 |
Gene Name: | family with sequence similarity 104, member A |
Gene Aliases: |
D11Wsu99e |
Location: |
Chr.11:113661319-113684186 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam104a | NM_138598.5 | 1 | 139 | NP_613064.2 |
AK009763.2 | BAB26487.1 | |||
AK017659.1 | BAB30858.1 | |||
AK028386.1 | BAC25923.1 | |||
AK087540.1 | BAC39919.1 | |||
AK145119.1 | ||||
AK159228.1 | BAE34914.1 | |||
AK159659.1 | BAE35268.1 | |||
AV572367.1 | 1 | 140 |