Assay Details
Target Gene Details
Entrez Gene ID: | 19157 |
Gene Name: | cytohesin 1 |
Gene Aliases: |
CLM1, CTH-1, CYTIP, Pscd1, mKIAA4240 |
Location: |
Chr.11:118164166-118248592 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyth1 | NM_001112699.1 | 13 | 2861 | NP_001106169.1 |
NM_001112700.1 | 14 | 2804 | NP_001106170.1 | |
NM_011180.3 | 14 | 2864 | NP_035310.2 | |
AA268675.1 | 1 | 94 | ||
AB035538.1 | 13 | 2833 | BAB13509.1 | |
AK210975.1 | ||||
AK220302.1 | 14 | 2786 | BAD90227.1 | |
AY126516.1 | 13 | 2830 | AAM95454.1 | |
BC057974.1 | 13 | 2745 | AAH57974.1 |
More Information
Set Membership: |
Intragenic Exonic |