Assay Details
Target Gene Details
Entrez Gene ID: | 30951 |
Gene Name: | chromobox 8 |
Gene Aliases: |
Pc3 |
Location: |
Chr.11:119038436-119040913 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cbx8 | NM_013926.1 | 5 | 673 | NP_038954.1 |
AB221673.1 | 5 | 610 | ||
AF180370.1 | 5 | 673 | AAF25615.1 | |
BC014815.1 | 5 | 623 | AAH14815.1 |
More Information
Set Membership: |
Intragenic Exonic |