Assay Details
Target Gene Details
Entrez Gene ID: | 194908 |
Gene Name: | phospholipase D family, member 6 |
Gene Aliases: |
4933433K01Rik, Gm10, Zuc, mZuc, mitoPLD |
Location: |
Chr.11:59783893-59787657 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pld6 | NM_001290283.1 | 2 | 656 | NP_001277212.1 |
NM_183139.2 | 2 | 652 | NP_898962.2 | |
NR_110894.1 | 2 | 407 | ||
AK077214.1 | 2 | 614 | BAC36687.1 | |
AK139586.1 | 2 | 640 | BAE24077.1 | |
CK137362.1 | 2 | 407 |
More Information
Set Membership: |
Intragenic Exonic |