Assay Details
Target Gene Details
Entrez Gene ID: | 18293 |
Gene Name: | oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide) |
Gene Aliases: |
2210403E04Rik, 2210412K19Rik, AA409584, d1401, mKIAA4192 |
Location: |
Chr.11:6291593-6359094 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ogdh | NM_001252282.1 | 24 | 4301 | NP_001239211.1 |
NM_001252283.1 | 24 | 4289 | NP_001239212.1 | |
NM_001252287.1 | 23 | 4332 | NP_001239216.1 | |
NM_001252288.1 | 23 | 4304 | NP_001239217.1 | |
NM_010956.4 | 23 | 4256 | NP_035086.2 | |
AK147289.1 | 23 | 4257 | BAE27824.1 |
More Information
Set Membership: |
Intragenic Exonic |