Assay Details
Target Gene Details
Entrez Gene ID: | 50997 |
Gene Name: | membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2) |
Gene Aliases: |
D11Bwg0652e, Dlg2, Dlgh2, Pals4 |
Location: |
Chr.11:102057016-102088636 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpp2 | NM_016695.3 | 13 | 2444 | NP_057904.1 |
AF162685.1 | 13 | 2431 | AAD44342.1 | |
AK141422.1 | 12 | 2442 | BAE24680.1 | |
AK161577.1 | 13 | 2437 | BAE36473.1 | |
AK172253.1 | 13 | 2429 | BAE42909.1 | |
BC053026.1 | 13 | 2444 | AAH53026.1 |
More Information
Set Membership: |
Intragenic Exonic |