Assay Details
Target Gene Details
Entrez Gene ID: | 217369 |
Gene Name: | urotensin 2 receptor |
Gene Aliases: |
Gpr14, UR-2-R, UTR, UTR2 |
Location: |
Chr.11:121160271-121161973 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uts2r | NM_145440.1 | 1 | 564 | NP_663415.1 |
AF441863.1 | 1 | 564 | AAL34551.1 | |
AY065981.1 | 1 | 523 | AAL55427.1 | |
BC125312.1 | 1 | 543 | ||
BC125318.1 | 1 | 543 |
More Information
Set Membership: |
Intragenic Exonic |