Assay Details
Target Gene Details
Entrez Gene ID: | 12330 |
Gene Name: | calnexin |
Gene Aliases: |
1110069N15Rik, AI988026, Cnx, D11Ertd153e |
Location: |
Chr.11:50293957-50325673 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Canx | NM_001110499.1 | 15 | 3894 | NP_001103969.1 |
NM_001110500.1 | 15 | 3862 | NP_001103970.1 | |
NM_007597.3 | 15 | 3898 | NP_031623.1 | |
AK017254.1 | 1 | 685 |
More Information
Set Membership: |
Intragenic Exonic |