Assay Details
Target Gene Details
Entrez Gene ID: | 70163 |
Gene Name: | LY6/PLAUR domain containing 8 |
Gene Aliases: |
2210415F13Rik |
Location: |
Chr.11:58379032-58390544 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lypd8 | NM_001083884.1 | 7 | 948 | NP_001077353.1 |
NM_027339.2 | 6 | 883 | NP_081615.2 | |
AK008940.1 | 7 | 937 | BAB25981.1 | |
AW121391.1 | 1 | 175 | ||
BC026828.1 | 6 | 858 | AAH26828.1 |
More Information
Set Membership: |
Intragenic Exonic |