Assay Details
Target Gene Details
Entrez Gene ID: | 68106 |
Gene Name: | 5'-nucleotidase, cytosolic IIIB |
Gene Aliases: |
2610037D24Rik, AI841843, C330027I04Rik, Nt5c3l |
Location: |
Chr.11:100422321-100441827 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nt5c3b | NM_001102650.1 | 9 | 1109 | NP_001096120.1 |
NM_026561.4 | NP_080837.3 | |||
AK006972.1 | BAB24814.1 | |||
AK082816.1 | BAC38632.1 | |||
AK148222.1 | 9 | 1085 | BAE28422.1 | |
AK162770.1 | 8 | 1187 | BAE37053.1 | |
BC015307.1 | 8 | 1064 | AAH15307.1 | |
BE944684.1 | 1 | 308 |
More Information
Set Membership: |
Intragenic Exonic |