Assay Details
Target Gene Details
Entrez Gene ID: | 52670 |
Gene Name: | cleavage and polyadenylation specific factor 4-like |
Gene Aliases: |
0610010C04Rik, 1500000C01Rik, D11Ertd636e |
Location: |
Chr.11:113696078-113711425 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cpsf4l | NM_001164532.1 | 8 | 1211 | NP_001158004.1 |
NM_026682.2 | 8 | 1055 | NP_080958.2 | |
NM_029794.2 | 7 | 1071 | NP_084070.1 | |
AK002449.1 | 7 | 958 | BAE43147.1 | |
AK005072.1 | 7 | 1071 | BAB23796.1 | |
BC016190.1 | 8 | 1049 | AAH16190.1 | |
DV657463.1 | 2 | 453 |
Target Gene Details
Entrez Gene ID: | 276852 |
Gene Name: | DNA segment, Chr 11, Wayne State University 47, expressed |
Gene Aliases: |
- |
Location: |
Chr.11:113684400-113727444 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
D11Wsu47e | AK139389.1 | BAE23989.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |