Assay Details
Target Gene Details
Entrez Gene ID: | 18612 |
Gene Name: | ets variant 4 |
Gene Aliases: |
AW414408, Pea-3, Pea3 |
Location: |
Chr.11:101769742-101785310 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Etv4 | NM_008815.2 | 13 | 1637 | NP_032841.2 |
AK144590.1 | 13 | 1590 | BAE25952.1 | |
AK144683.1 | 11 | 1353 | ||
AK166136.1 | 13 | 1573 | ||
DQ832277.1 | 12 | 1342 |
More Information
Set Membership: |
Intragenic Exonic |