Assay Details
Target Gene Details
Entrez Gene ID: | 68832 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
1110057K04Rik |
Location: |
Chr.12:8208100-8299566 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ldah | NM_001167767.1 | 6 | 1716 | NP_001161239.1 |
NM_172401.4 | 7 | 1807 | NP_765989.3 | |
AK004285.1 | 1 | 393 | ||
AK076460.1 | 5 | 1473 | BAC36352.1 | |
AK079099.1 | 7 | 1791 | BAC37539.1 | |
BC046986.1 | 6 | 1726 | AAH46986.1 | |
BC057311.1 | 6 | 1696 | AAH57311.1 |
More Information
Set Membership: |
Intragenic Exonic |