Assay Details
Target Gene Details
Entrez Gene ID: | 52898 |
Gene Name: | ribonuclease, RNase K |
Gene Aliases: |
2310033H11Rik, AA571397, D11Bwg0434e |
Location: |
Chr.11:70238123-70239852 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnasek | NM_173742.3 | 3 | 448 | NP_776103.1 |
AK003548.1 | 3 | 424 | ||
AK009592.1 | 3 | 443 | BAC25261.1 | |
AK018709.1 | 3 | 397 | ||
AK171975.1 | 3 | 397 | BAE42755.1 | |
BB308989.1 | 1 | 103 | ||
BC021602.1 | 3 | 376 | AAH21602.2 | |
BC021603.1 | 3 | 376 | AAH21603.2 | |
BC024417.1 | 3 | 406 | AAH24417.1 |