Assay Details
Target Gene Details
Entrez Gene ID: | 57837 |
Gene Name: | Era (G-protein)-like 1 (E. coli) |
Gene Aliases: |
2610524P08Rik, 9130407C09Rik, AU019798, Era, M-ERA, MERA-S, MERA-W |
Location: |
Chr.11:78073376-78080383 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eral1 | NM_022313.2 | 10 | 1350 | NP_071708.2 |
AB049389.1 | 10 | 1326 | BAB56113.1 | |
AF190730.1 | 10 | 1318 | AAG17176.1 | |
AF190731.1 | 9 | 1086 | AAG17177.1 | |
AK012155.1 | 10 | 1340 | BAB28065.1 | |
AK153947.1 | 2 | 1831 | ||
BC019728.1 | 4 | 538 | AAH19728.1 | |
BC068271.1 | 10 | 1345 | AAH68271.1 |
More Information
Set Membership: |
Intragenic Exonic |