Assay Details
Target Gene Details
Entrez Gene ID: | 74230 |
Gene Name: | RIKEN cDNA 1700016K19 gene |
Gene Aliases: |
Liat1 |
Location: |
Chr.11:75999912-76003569 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700016K19Rik | NM_198637.2 | 1 | 59 | NP_941039.1 |
AK006044.1 | ||||
AK132806.1 | BAE21367.1 | |||
BC049758.1 | AAH49758.1 | |||
BQ931096.1 | 1 | 74 |