Assay Details
Target Gene Details
Entrez Gene ID: | 14615 |
Gene Name: | gap junction protein, gamma 1 |
Gene Aliases: |
C130009G16Rik, Cnx45, Cx45, Gja-7, Gja7 |
Location: |
Chr.11:102799472-102819686 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gjc1 | NM_001159382.1 | 3 | 564 | NP_001152854.1 |
NM_001159383.1 | 3 | 557 | NP_001152855.1 | |
NM_008122.3 | 3 | 1113 | NP_032148.1 | |
AK081351.1 | 2 | 3245 | BAC38203.1 | |
BC050840.1 | 3 | 564 | AAH50840.1 | |
BC071230.1 | 3 | 557 | AAH71230.1 | |
X63100.1 | 3 | 734 | CAA44814.1 |
More Information
Set Membership: |
Intragenic Exonic |