Assay Details
Target Gene Details
Entrez Gene ID: | 53321 |
Gene Name: | contactin associated protein-like 1 |
Gene Aliases: |
AI841080, Caspr, NCP1, Nrxn4, p190, shm |
Location: |
Chr.11:101174629-101190727 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntnap1 | NM_016782.2 | 8 | 1269 | NP_058062.2 |
AF039833.1 | 8 | 1279 | AAB96760.1 |