Assay Details
Target Gene Details
Entrez Gene ID: | 20926 |
Gene Name: | suppressor of Ty 6 |
Gene Aliases: |
5131400N11Rik, AI132449, SPT6, Supt6h, mKIAA0162 |
Location: |
Chr.11:78206749-78245703 on Build GRCm38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supt6 | NM_009297.2 | 37 | 5697 | NP_033323.2 |
AK046156.1 | 8 | 1438 | BAC32616.1 | |
AK129069.1 | 25 | 4020 | BAC97879.1 | |
AK158846.1 | 19 | 3632 | BAE34691.1 | |
AK207208.1 | 1 | 148 | ||
BC072657.1 | 37 | 5617 | AAH72657.1 | |
U40375.1 | 37 | 5695 | AAB18950.1 |
More Information
Set Membership: |
Intragenic Exonic |