Assay Details
Target Gene Details
Entrez Gene ID: | 22361 |
Gene Name: | vanin 1 |
Gene Aliases: |
V-1 |
Location: |
Chr.10:23894688-23905344 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vnn1 | NM_011704.3 | 5 | 1136 | NP_035834.2 |
AJ132098.1 | 5 | 969 | CAA10567.1 | |
AK002773.1 | 5 | 968 | BAB22347.1 | |
AK145984.1 | 5 | 1042 | BAE26806.1 | |
AK149485.1 | 3 | 517 | BAE28911.1 | |
AK167427.1 | 5 | 968 | BAE39515.1 | |
BC019203.1 | 5 | 962 | AAH19203.1 |
More Information
Set Membership: |
Intragenic Exonic |