Assay Details
Target Gene Details
Entrez Gene ID: | 13649 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
Location: |
Chr.11:16752203-16913907 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egfr | NM_007912.4 | 1 | 249 | NP_031938.1 |
NM_207655.2 | 1 | 249 | NP_997538.1 | |
AF124513.1 | 1 | 184 | AAD44149.1 | |
AF275367.1 | 1 | 193 | AAG24386.1 | |
AK004883.1 | 1 | 250 | BAB23641.1 | |
AK004911.1 | 1 | 224 | BAB23662.1 | |
AK004944.1 | 1 | 243 | BAB23688.1 | |
AK014017.1 | 1 | 223 | ||
AK049452.1 | 1 | 111 | ||
AK134262.1 | 1 | 223 | ||
BC023729.1 | 1 | 220 | AAH23729.1 | |
U03425.1 | AAA17899.1 | |||
X59698.1 | 1 | 128 | CAA42219.1 | |
X78987.1 | CAA55587.1 |
More Information
Set Membership: |
Intragenic Exonic |