Assay Details
Target Gene Details
Entrez Gene ID: | 66102 |
Gene Name: | chemokine (C-X-C motif) ligand 16 |
Gene Aliases: |
0910001K24Rik, AV290116, BB024863, CXCL16v1, CXCL16v2, SR-PSOX, Zmynd15, b2b498Clo |
Location: |
Chr.11:70454234-70459984 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl16 | NM_023158.6 | NP_075647.3 | ||
AK028875.1 | BAC26166.1 |
Target Gene Details
Entrez Gene ID: | 574428 |
Gene Name: | zinc finger, MYND-type containing 15 |
Gene Aliases: |
- |
Location: |
Chr.11:70459622-70466207 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zmynd15 | NM_001029929.2 | 1 | 377 | NP_001025100.1 |
AK029988.1 | 1 | 378 | BAC26719.1 | |
AK171931.1 | 1 | 570 | BAE42737.1 | |
BC151086.1 | 1 | 49 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |