Assay Details
Target Gene Details
Entrez Gene ID: | 217154 |
Gene Name: | SH3 and cysteine rich domain 2 |
Gene Aliases: |
24b2, 24b2/STAC2, AW240854 |
Location: |
Chr.11:98036624-98053462 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stac2 | NM_146028.4 | 1 | 486 | NP_666140.1 |
AJ608761.1 | 1 | 299 | CAE75538.1 | |
AK078132.1 | 1 | 487 | BAC37142.1 | |
AK079357.1 | 1 | 345 | BAC37621.1 | |
AK144433.1 | 1 | 342 | BAE25886.1 | |
BC024864.1 | 1 | 392 | AAH24864.1 | |
BC029794.1 | 1 | 333 | AAH29794.1 |