Assay Details
Target Gene Details
Entrez Gene ID: | 14799 |
Gene Name: | glutamate receptor, ionotropic, AMPA1 (alpha 1) |
Gene Aliases: |
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1 |
Location: |
Chr.11:57011571-57330244 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gria1 | NM_001113325.2 | 16 | 5265 | NP_001106796.1 |
NM_001252403.1 | 16 | 4892 | NP_001239332.1 | |
NM_008165.4 | 16 | 5265 | NP_032191.2 | |
AK013661.1 | 1 | 701 | ||
AK135107.1 | 16 | 4893 | ||
BC056397.1 | 16 | 4927 | AAH56397.1 | |
BC060702.1 | 16 | 4848 | AAH60702.1 | |
BQ176734.1 | 1 | 491 |
More Information
Set Membership: |
Intragenic Exonic |