Assay Details
Target Gene Details
Entrez Gene ID: | 320398 |
Gene Name: | leucine-rich repeats and immunoglobulin-like domains 3 |
Gene Aliases: |
9030421L11Rik, 9130004I02Rik, 9430095K15Rik, mKIAA3016 |
Location: |
Chr.10:125966219-126015359 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrig3 | NM_177152.5 | 18 | 3224 | NP_796126.4 |
AK018522.1 | 12 | 2037 | ||
AK020260.1 | 2 | 1652 | ||
AK078783.1 | 4 | 818 | BAC37394.1 | |
AK129481.1 | 14 | 2173 | BAC98291.1 | |
AK135388.1 | 2 | 856 | BAE22516.1 | |
AK214999.1 | 1 | 76 | ||
AY505341.1 | 18 | 3167 | AAR98630.1 | |
BC065142.1 | 18 | 3190 | AAH65142.1 |
More Information
Set Membership: |
Intragenic Exonic |