Assay Details
Target Gene Details
Entrez Gene ID: | 58250 |
Gene Name: | carbohydrate sulfotransferase 11 |
Gene Aliases: |
1110020P09Rik, C4ST, C4ST-1, C4ST1, C4s |
Location: |
Chr.10:82985380-83195907 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chst11 | NM_021439.2 | 3 | 1398 | NP_067414.2 |
AB030378.1 | BAA92942.1 | |||
AJ289133.1 | 3 | 1406 | CAB92141.1 | |
BC137629.1 | 3 | 1146 | ||
BC144793.1 | 3 | 1131 |
More Information
Set Membership: |
Intragenic Exonic |