Assay Details
Target Gene Details
Entrez Gene ID: | 67968 |
Gene Name: | oocyte expressed protein |
Gene Aliases: |
2410146L05Rik, Floped, Moep19, Sddr |
Location: |
Chr.9:78376903-78378588 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ooep | NM_026480.3 | 3 | 563 | NP_080756.1 |
AB283026.1 | ||||
AK010266.1 | 3 | 558 | ||
AK010809.1 | 3 | 564 | BAB27195.1 | |
AK133554.1 | 3 | 528 | BAE21721.1 | |
AK135950.1 | 3 | 529 | BAE22739.1 | |
AK145510.1 | 3 | 529 | BAE26478.1 | |
BC062245.1 | 3 | 514 | AAH62245.1 | |
DQ238107.1 | 3 | 564 | ABB30176.1 |