Assay Details
Target Gene Details
Entrez Gene ID: | 104360 |
Gene Name: | insulin related protein 2 (islet 2) |
Gene Aliases: |
3110001N10Rik |
Location: |
Chr.9:55538677-55547667 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Isl2 | NM_027397.3 | 4 | 697 | NP_081673.2 |
AK013964.1 | 4 | 640 | BAB29088.1 | |
BC120880.2 | 3 | 377 | ||
BC148307.2 | 3 | 377 |