Assay Details
Target Gene Details
Entrez Gene ID: | 16798 |
Gene Name: | large tumor suppressor |
Gene Aliases: |
AW208599 |
Location: |
Chr.10:7681209-7716461 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lats1 | NM_010690.1 | 8 | 6558 | NP_034820.1 |
AK029530.1 | 1 | 2009 | ||
AK047022.1 | 1 | 1249 | ||
AK081925.1 | 1 | 1795 | ||
AK151387.1 | 1 | 2224 |
More Information
Set Membership: |
Intragenic Exonic |