Assay Details
Target Gene Details
Entrez Gene ID: | 378431 |
Gene Name: | taxilin beta |
Gene Aliases: |
2310001N14Rik, Mdp77 |
Location: |
Chr.10:17796091-17845665 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Txlnb | NM_138628.3 | 10 | 2973 | NP_619534.2 |
AF422245.1 | 10 | 2910 | AAL33910.1 | |
AK137442.1 | 10 | 2888 | BAE23352.1 | |
BC141023.1 |
More Information
Set Membership: |
Intragenic Exonic |