Assay Details
Target Gene Details
Entrez Gene ID: | 20669 |
Gene Name: | SRY (sex determining region Y)-box 14 |
Gene Aliases: |
- |
Location: |
Chr.9:99874106-99876170 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox14 | NM_011440.1 | 1 | 1376 | NP_035570.1 |
AF193435.1 | 1 | 875 | AAF62396.1 | |
AK138208.1 | 1 | 1268 | BAE23582.1 | |
BC044735.2 | 2 | 734 | AAH44735.1 | |
BC055767.1 | 1 | 1366 | AAH55767.1 | |
BC069948.1 | 1 | 1266 | AAH69948.1 | |
BC083360.1 | 1 | 1373 | AAH83360.1 | |
BC100555.1 | 1 | 1376 | AAI00556.1 |
More Information
Set Membership: |
Intragenic Exonic |