Assay Details
Target Gene Details
Entrez Gene ID: | 17311 |
Gene Name: | kit ligand |
Gene Aliases: |
Clo, Con, Gb, Kitlg, Mgf, SCF, SF, SLF, Sl, blz, contrasted |
Location: |
Chr.10:100015809-100100416 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kitl | NM_013598.2 | 10 | 1793 | NP_038626.1 |
AK018777.1 | 10 | 1793 | BAB31402.1 | |
AK088655.1 | 1 | 675 | ||
AK134301.1 | 10 | 1793 | BAE22091.1 | |
AK165233.1 | 10 | 1791 | BAE38091.1 | |
BC011322.1 | 10 | 1763 | AAH11322.1 | |
M57647.1 | 10 | 1745 | AAA39538.1 | |
U44725.1 | 10 | 1793 | AAC52447.1 |