Assay Details
Target Gene Details
Entrez Gene ID: | 58802 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 4 |
Gene Aliases: |
1700058G18Rik, 2900045G12Rik |
Location: |
Chr.10:116417868-116473882 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnmb4 | NM_021452.1 | 1 | 116 | NP_067427.1 |
AF215892.1 | 1 | 116 | AAF75597.1 | |
AK013647.1 | 1 | 476 | ||
AK050596.1 | ||||
BC117779.2 | 1 | 116 |
More Information
Set Membership: |
Intragenic Exonic |