Assay Details
Target Gene Details
Entrez Gene ID: | 13713 |
Gene Name: | ELK3, member of ETS oncogene family |
Gene Aliases: |
D430049E23Rik, Erp, Etrp, Net, Sap-2 |
Location: |
Chr.10:93247415-93311167 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elk3 | NM_001282967.1 | 1 | 204 | NP_001269896.1 |
NM_013508.2 | 1 | 204 | NP_038536.2 | |
AK088129.1 | ||||
BC005686.1 | 1 | 180 | AAH05686.1 | |
BC053402.1 | AAH53402.1 | |||
BC055735.1 | AAH55735.1 | |||
L19953.1 | 1 | 95 | AAA19872.1 | |
Z32815.1 | 1 | 84 | CAA83676.1 |
More Information
Set Membership: |
Intragenic Exonic |