Assay Details
Target Gene Details
Entrez Gene ID: | 18674 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 3 |
Gene Aliases: |
5730556H19Rik, PTP, Phc |
Location: |
Chr.10:91116578-91124052 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc25a3 | NM_133668.3 | 8 | 1425 | NP_598429.1 |
AK077274.1 | 8 | 1426 | BAC36723.1 | |
AK151887.1 | 8 | 1430 | BAE30772.1 |
More Information
Set Membership: |
Intragenic Exonic |