Assay Details
Target Gene Details
Entrez Gene ID: | 27403 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 7 |
Gene Aliases: |
ABCX, Abc51 |
Location: |
Chr.10:79996453-80015572 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abca7 | NM_013850.1 | 29 | 4058 | NP_038878.1 |
AF287141.1 | 29 | 4058 | AAK56862.1 | |
AK143653.1 | 4 | 544 | ||
BC145120.1 | 29 | 4121 | ||
BC150708.1 | 29 | 4121 |
More Information
Set Membership: |
Intragenic Exonic |