Assay Details
Target Gene Details
Entrez Gene ID: | 93759 |
Gene Name: | sirtuin 1 |
Gene Aliases: |
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha |
Location: |
Chr.10:63319005-63339061 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt1 | NM_001159589.1 | 8 | 2823 | NP_001153061.1 |
NM_019812.2 | 9 | 2937 | NP_062786.1 | |
AF214646.1 | 9 | 2922 | AAF24983.1 | |
AK141245.1 | 1 | 715 | ||
AK144203.1 | 8 | 2826 | BAE25766.1 | |
AY377984.1 | 9 | 2922 | AAR23928.1 | |
BC006584.1 | 2 | 1243 | AAH06584.1 |
More Information
Set Membership: |
Intragenic Exonic |