Assay Details
Target Gene Details
Entrez Gene ID: | 18483 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.10:79793351-79820896 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Palm | NM_001161747.1 | 8 | 1280 | NP_001155219.1 |
NM_023128.4 | 9 | 1412 | NP_075617.3 | |
AK009200.1 | 7 | 1121 | ||
AK037044.1 | 9 | 1269 | BAC29683.1 | |
AK082163.1 | 8 | 1282 | BAC38429.1 | |
AK129107.1 | 7 | 1151 | BAC97917.1 | |
AK162411.1 | 8 | 1273 | BAE36902.1 | |
AK185384.1 | ||||
BC015297.1 | 8 | 1264 | AAH15297.1 | |
Y14771.1 | 9 | 1375 | CAB37403.1 |