Assay Details
Target Gene Details
Entrez Gene ID: | 102423 |
Gene Name: | histone H4 transcription factor |
Gene Aliases: |
AA589481, HiNF-P, Mizf |
Location: |
Chr.9:44292438-44305705 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hinfp | NM_172162.3 | 10 | 1646 | NP_751894.2 |
AK049348.1 | 10 | 1608 | ||
BC029091.1 | 9 | 1629 | AAH29091.1 |
More Information
Set Membership: |
Intragenic Exonic |