Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc41 | NM_001003910.2 | 2 | 361 | NP_001003910.2 |
NM_001003939.2 | 3 | 470 | NP_001003939.2 | |
NM_153595.3 | 1 | 154 | NP_705823.3 | |
AK133195.1 | 2 | 301 | ||
AY651019.1 | 4 | 553 | AAT81231.1 | |
AY651020.1 | AAT81232.1 | |||
AY651021.1 | AAT81233.1 | |||
BC030307.1 | 3 | 448 | AAH30307.1 |