Assay Details
Target Gene Details
Entrez Gene ID: | 27375 |
Gene Name: | tight junction protein 3 |
Gene Aliases: |
- |
Location: |
Chr.10:81273188-81291581 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tjp3 | NM_001282095.1 | 5 | 613 | NP_001269024.1 |
NM_001282096.1 | 5 | 676 | NP_001269025.1 | |
NM_013769.3 | 5 | 715 | NP_038797.2 | |
AF157006.1 | 5 | 683 | AAF24175.1 | |
AK029083.1 | 5 | 662 | BAC26285.1 | |
AK134100.1 | 5 | 716 | BAE22013.1 | |
AK167587.1 | 5 | 614 | ||
BC012518.1 | 5 | 680 | AAH12518.1 |
More Information
Set Membership: |
Intragenic Exonic |