Assay Details
Target Gene Details
Entrez Gene ID: | 13139 |
Gene Name: | diacylglycerol kinase, alpha |
Gene Aliases: |
80kDa, AW146112, Dagk1 |
Location: |
Chr.10:128720134-128748420 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dgka | NM_016811.2 | 1 | 91 | NP_058091.2 |
AK041812.1 | 1 | 917 | ||
BC006713.1 | 1 | 91 | AAH06713.1 | |
JN832677.1 | 2 | 123 | ||
JN832678.1 | 1 | 110 | ||
JN832679.1 | 1 | 110 | ||
JN832681.1 | 2 | 123 | ||
JN832683.1 | 1 | 110 |
More Information
Set Membership: |
Intragenic Exonic |