Assay Details
Target Gene Details
Entrez Gene ID: | 13202 |
Gene Name: | D-dopachrome tautomerase |
Gene Aliases: |
C78655 |
Location: |
Chr.10:75771233-75773374 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddt | NM_010027.1 | 3 | 383 | NP_034157.1 |
AK144193.1 | 3 | 426 | BAE25759.1 | |
BC010753.1 | 3 | 377 | AAH10753.1 |
More Information
Set Membership: |
Intragenic Exonic |