Assay Details
Target Gene Details
Entrez Gene ID: | 67331 |
Gene Name: | ATPase, class I, type 8B, member 3 |
Gene Aliases: |
1700042F02Rik, 1700056N23Rik, ATPIK, SAPLT |
Location: |
Chr.10:80519584-80539150 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp8b3 | NM_026094.3 | 22 | 2409 | NP_080370.2 |
AY364445.1 | 22 | 2409 | AAR12913.1 | |
BC118977.1 | 22 | 2409 | ||
BC118978.1 |
More Information
Set Membership: |
Intragenic Exonic |