Assay Details
Target Gene Details
Entrez Gene ID: | 66043 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit |
Gene Aliases: |
0610008F14Rik, 1500000I11Rik, AA960090, AI876556, AU020773, C85518 |
Location: |
Chr.10:80142280-80145818 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atp5d | NM_025313.2 | 4 | 605 | NP_079589.2 |
AK002330.1 | 4 | 555 | BAB22016.1 | |
AK005082.1 | 4 | 549 | ||
AK011374.1 | 4 | 526 | BAB27577.1 | |
AK018011.1 | 4 | 551 | BAB31035.1 | |
AK076064.1 | 4 | 526 | ||
AK151151.1 | 4 | 540 | BAE30157.1 | |
AK155285.1 | 4 | 589 | BAE33165.1 | |
AK165932.1 | 4 | 554 | BAE38468.1 | |
AK206431.1 | ||||
BC008273.1 | 4 | 514 | AAH08273.1 | |
BY451544.1 | 2 | 140 | ||
CT010177.1 | 4 | 454 | CAJ18385.1 |
More Information
Set Membership: |
Intragenic Exonic |